MAN326367037 Manchester Organics

2-Chloro-3-(trifluoromethyl)quinoline

  • MDL number MFCD08741354
  • Cas Number : 25199-86-4
  • Molecular Weight : 231.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-3-(trifluoromethyl)quinoline
Inchi Code 1S/C10H5ClF3N/c11-9-7(10(12,13)14)5-6-3-1-2-4-8(6)15-9/h1-5H
Inchi Key UHUURLGTUPTFDC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/R14630
Availability details for
Quantity
  • Availability
  • Add to Cart