MAN326367038 Manchester Organics

3-Bromo-2-fluoro-6-methylpyridine

  • MDL number MFCD03095080
  • Cas Number : 375368-78-8
  • Molecular Weight : 190.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/K21867
Availability details for
Quantity
  • Availability
  • Add to Cart