MAN326367074 Manchester Organics

2-(Difluoromethyl)-6-methylpyridine

  • Molecular Weight : 143.134

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N14651
Availability details for
Quantity
  • Availability
  • Add to Cart