MAN326367085 Manchester Organics

3-Amino-4-ethylpyrazole oxalate

  • MDL number MFCD11114053
  • Molecular Weight : 201.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A21904

Safety Information

MSDS https://www.manchesterorganics.com/A21904
Availability details for
Quantity
  • Availability
  • Add to Cart