MAN326367090 Manchester Organics

Ethyl morpholine-3-carboxylate hydrochloride

  • MDL number MFCD12755498
  • Cas Number : 84005-98-1
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-morpholinecarboxylate hydrochloride
Inchi Code 1S/C7H13NO3.ClH/c1-2-11-7(9)6-5-10-4-3-8-6;/h6,8H,2-5H2,1H3;1H
Inchi Key RERWECVXIGGYMH-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin GB
Product Link https://www.manchesterorganics.com/R11032
Availability details for
Quantity
  • Availability
  • Add to Cart