MAN326367099 Manchester Organics

3-Fluoro-2-methoxybenzonitrile

  • MDL number MFCD06637312
  • Molecular Weight : 151.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M21892
Availability details for
Quantity
  • Availability
  • Add to Cart