MAN326367108 Manchester Organics

3-Bromo-5(methylsulfonyl)pyridine

  • MDL number MFCD09801057
  • Molecular Weight : 236.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W22303

Safety Information

MSDS https://www.manchesterorganics.com/W22303
Availability details for
Quantity
  • Availability
  • Add to Cart