MAN326367122 Manchester Organics

2-(Difluoromethyl)-4-fluoro-1-nitrobenzene

  • MDL number MFCD14698664
  • Cas Number : 1214333-17-1
  • Molecular Weight : 191.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/F14667
Availability details for
Quantity
  • Availability
  • Add to Cart