MAN326367132 Manchester Organics

2-Chloro-4-(difluoromethyl)-1-fluorobenzene

  • MDL number MFCD14698545
  • Cas Number : 1214372-57-2
  • Molecular Weight : 180.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4-(difluoromethyl)-1-fluorobenzene
Inchi Code 1S/C7H4ClF3/c8-5-3-4(7(10)11)1-2-6(5)9/h1-3,7H
Inchi Key XEHBLRGJWPBARX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/S14677
Availability details for
Quantity
  • Availability
  • Add to Cart