MAN326367139 Manchester Organics

4-(Difluoromethyl)-2-fluoro-1-methylbenzene

  • MDL number MFCD14698598
  • Cas Number : 1214334-21-0
  • Molecular Weight : 160.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(difluoromethyl)-2-fluoro-1-methylbenzene
Inchi Code 1S/C8H7F3/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4,8H,1H3
Inchi Key UCOCOWUDEBTMBY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/T14678
Availability details for
Quantity
  • Availability
  • Add to Cart