MAN326367153 Manchester Organics

5-Chloro-2-fluoro-3-methylpyridine

  • MDL number MFCD03095094
  • Molecular Weight : 145.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y22399

Safety Information

MSDS https://www.manchesterorganics.com/Y22399
Availability details for
Quantity
  • Availability
  • Add to Cart