MAN326367173 Manchester Organics

3-(Trifluoromethoxy)phenyl-DL-alanine

  • MDL number MFCD04116035
  • Molecular Weight : 249.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H12090

Safety Information

MSDS https://www.manchesterorganics.com/H12090
Availability details for
Quantity
  • Availability
  • Add to Cart