MAN326367174 Manchester Organics

4-(Trifluoromethoxy)phenyl-DL-alanine

  • MDL number MFCD00764461
  • Molecular Weight : 249.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G12089

Safety Information

MSDS https://www.manchesterorganics.com/G12089
Availability details for
Quantity
  • Availability
  • Add to Cart