MAN326367180 Manchester Organics

2-Bromophenyl-DL-alanine

  • MDL number No data
  • Molecular Weight : 244.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P12190

Safety Information

MSDS https://www.manchesterorganics.com/P12190
Availability details for
Quantity
  • Availability
  • Add to Cart