MAN326367208 Manchester Organics

4-Bromo-3-chloropyridine

  • MDL number MFCD04112577
  • Molecular Weight : 192.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N21964
Availability details for
Quantity
  • Availability
  • Add to Cart