MAN326367229 Manchester Organics

5-Chloro-2-(trifluoromethyl)phenylacetic acid

  • MDL number MFCD01631358
  • Molecular Weight : 238.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R22156

Safety Information

MSDS https://www.manchesterorganics.com/R22156
Availability details for
Quantity
  • Availability
  • Add to Cart