MAN326367273 Manchester Organics

2-Fluoro-6-(trifluoromethyl)phenylacetic acid

  • MDL number MFCD00061191
  • Molecular Weight : 222.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F22099

Safety Information

MSDS https://www.manchesterorganics.com/F22099
Availability details for
Quantity
  • Availability
  • Add to Cart