MAN326367285 Manchester Organics

2,3,6-Tribromopyridine

  • MDL number MFCD00234000
  • Cas Number : 2402-92-8
  • Molecular Weight : 315.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/U22040
Availability details for
Quantity
  • Availability
  • Add to Cart