MAN326367291 Manchester Organics

2,3,4-Trifluorophenylacetic acid

  • MDL number MFCD00061235
  • Molecular Weight : 190.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B22072

Safety Information

MSDS https://www.manchesterorganics.com/B22072
Availability details for
Quantity
  • Availability
  • Add to Cart