MAN326367304 Manchester Organics

2,6-Difluoro-3-methylbenzoic acid

  • MDL number MFCD01631332
  • Molecular Weight : 172.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R22227

Safety Information

MSDS https://www.manchesterorganics.com/R22227
Availability details for
Quantity
  • Availability
  • Add to Cart