MAN326367334 Manchester Organics

3-(3,4-Dimethylphenyl)propionic acid

  • MDL number MFCD03425686
  • Molecular Weight : 178.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W22491

Safety Information

MSDS https://www.manchesterorganics.com/W22491
Availability details for
Quantity
  • Availability
  • Add to Cart