MAN326367336 Manchester Organics

4-Isopropylphenylacetic acid

  • MDL number MFCD00041035
  • Molecular Weight : 178.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y22541

Safety Information

MSDS https://www.manchesterorganics.com/Y22541
Availability details for
Quantity
  • Availability
  • Add to Cart