MAN326367337 Manchester Organics

3-(4-Isopropylphenyl)propionic acid

  • MDL number MFCD01462077
  • Molecular Weight : 192.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z22613

Safety Information

MSDS https://www.manchesterorganics.com/Z22613
Availability details for
Quantity
  • Availability
  • Add to Cart