MAN326367344 Manchester Organics

2-Methyl-5-bromopyrimidine

  • MDL number MFCD07375143
  • Cas Number : 7752-78-5
  • Molecular Weight : 173.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/G22100
Availability details for
Quantity
  • Availability
  • Add to Cart