MAN326367346 Manchester Organics

5-Bromo-2-fluorophenylacetic acid

  • MDL number MFCD06246020
  • Cas Number : 883514-21-4
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/J22102
Availability details for
Quantity
  • Availability
  • Add to Cart