MAN326367348 Manchester Organics

2-Chloro-5-fluorophenylacetic acid

  • MDL number MFCD01016155
  • Molecular Weight : 188.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L22246

Safety Information

MSDS https://www.manchesterorganics.com/L22246
Availability details for
Quantity
  • Availability
  • Add to Cart