MAN326367354 Manchester Organics

5-Chloro-2,4-difluoroaniline

  • MDL number MFCD06808806
  • Molecular Weight : 163.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V22444

Safety Information

MSDS https://www.manchesterorganics.com/V22444
Availability details for
Quantity
  • Availability
  • Add to Cart