MAN326367361 Manchester Organics

N-Boc-Piperidine-4-carboxylic acid methyl ester

  • MDL number MFCD02183584
  • Molecular Weight : 243.299

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D22120
Availability details for
Quantity
  • Availability
  • Add to Cart