MAN326367386 Manchester Organics

5-Ethylthiophene-3-carboxylic acid

  • MDL number MFCD03419849
  • Molecular Weight : 156.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D22216

Safety Information

MSDS https://www.manchesterorganics.com/D22216
Availability details for
Quantity
  • Availability
  • Add to Cart