MAN326367391 Manchester Organics

1-(Tetrahydropyranyl)-1H-indazole-5-carboxylic acid

  • MDL number MFCD09870060
  • Molecular Weight : 246.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J22221

Safety Information

MSDS https://www.manchesterorganics.com/J22221
Availability details for
Quantity
  • Availability
  • Add to Cart