MAN326367394 Manchester Organics

2-Methyl-2H-indazole-6-boronic acid pinacol ester

  • MDL number MFCD11109435
  • Molecular Weight : 258.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N22319

Safety Information

MSDS https://www.manchesterorganics.com/N22319
Availability details for
Quantity
  • Availability
  • Add to Cart