MAN326367404 Manchester Organics

7-Methyl-1H-indazole-5-boronic acid

  • MDL number MFCD12028646
  • Molecular Weight : 175.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B22212

Safety Information

MSDS https://www.manchesterorganics.com/B22212
Availability details for
Quantity
  • Availability
  • Add to Cart