MAN326367410 Manchester Organics

7-Bromoindazole-1-carboxylic acid tert-butyl ester

  • MDL number MFCD11505878
  • Molecular Weight : 297.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H22243

Safety Information

MSDS https://www.manchesterorganics.com/H22243
Availability details for
Quantity
  • Availability
  • Add to Cart