MAN326367417 Manchester Organics

4-Amino-3-bromo-1-methylindazole

  • MDL number MFCD11109355
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R22344

Safety Information

MSDS https://www.manchesterorganics.com/R22344
Availability details for
Quantity
  • Availability
  • Add to Cart