MAN326367425 Manchester Organics

5-Bromo-1-methylindazole

  • MDL number MFCD09878568
  • Molecular Weight : 211.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z22707

Safety Information

MSDS https://www.manchesterorganics.com/Z22707
Availability details for
Quantity
  • Availability
  • Add to Cart