MAN326367450 Manchester Organics

6-Formyl-1-methyl-1H-indazole

  • MDL number MFCD11109366
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B22237

Safety Information

MSDS https://www.manchesterorganics.com/B22237
Availability details for
Quantity
  • Availability
  • Add to Cart