MAN326367451 Manchester Organics

5-Formyl-1-methyl-1H-indazole

  • MDL number MFCD11109365
  • Molecular Weight : 160.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A22236

Safety Information

MSDS https://www.manchesterorganics.com/A22236
Availability details for
Quantity
  • Availability
  • Add to Cart