MAN326367473 Manchester Organics

1-N-BOC-piperazine-2-carboxylic acid methyl ester

  • MDL number MFCD01861779
  • Cas Number : 129799-15-1
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/C22238
Availability details for
Quantity
  • Availability
  • Add to Cart