MAN326367475 Manchester Organics

(R)-4-N-Boc-piperazine-2-carboxylic acid methyl ester

  • MDL number MFCD04115325
  • Molecular Weight : 244.287

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E22240
Availability details for
Quantity
  • Availability
  • Add to Cart