MAN326367480 Manchester Organics

1-Boc-2-ethyl-piperazine

  • MDL number MFCD03265490
  • Cas Number : 393781-71-0
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Country of Origin GB
Product Link https://www.manchesterorganics.com/K22245
Availability details for
Quantity
  • Availability
  • Add to Cart