MAN326367485 Manchester Organics

1-Boc -2-propyl-piperazine

  • MDL number MFCD08273955
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N22413

Safety Information

MSDS https://www.manchesterorganics.com/N22413
Availability details for
Quantity
  • Availability
  • Add to Cart