MAN326367488 Manchester Organics

(S)-1-Boc-3-ethyl-piperazine

  • MDL number MFCD07772110
  • Molecular Weight : 214.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T22488

Safety Information

MSDS https://www.manchesterorganics.com/T22488
Availability details for
Quantity
  • Availability
  • Add to Cart