MAN326367491 Manchester Organics

1-Boc-3-propyl-piperazine

  • MDL number MFCD08273956
  • Cas Number : 502649-27-6
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/W22255
Availability details for
Quantity
  • Availability
  • Add to Cart