MAN326367493 Manchester Organics

(R)-1-Boc-3-isopropyl-piperazine

  • MDL number MFCD07772113
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y22706

Safety Information

MSDS https://www.manchesterorganics.com/Y22706
Availability details for
Quantity
  • Availability
  • Add to Cart