MAN326367494 Manchester Organics

1-Boc-3-isopropyl-piperazine

  • MDL number MFCD08273958
  • Cas Number : 502649-32-3
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-isopropyl-1-piperazinecarboxylate
Inchi Code 1S/C12H24N2O2/c1-9(2)10-8-14(7-6-13-10)11(15)16-12(3,4)5/h9-10,13H,6-8H2,1-5H3
Inchi Key UHLAQCKNCBYTIF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z22258
Availability details for
Quantity
  • Availability
  • Add to Cart