MAN326367495 Manchester Organics

1-Boc-2-butyl-piperazine

  • MDL number MFCD08273959
  • Molecular Weight : 242.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B22285

Safety Information

MSDS https://www.manchesterorganics.com/B22285
Availability details for
Quantity
  • Availability
  • Add to Cart