MAN326367499 Manchester Organics

(R)-1-Boc-2-Hydroxymethyl-piperazine

  • MDL number MFCD07772092
  • Molecular Weight : 216.277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G22265
Availability details for
Quantity
  • Availability
  • Add to Cart