MAN326367500 Manchester Organics

1-Boc-2-(hydroxymethyl)piperazine

  • MDL number MFCD07772091
  • Molecular Weight : 216.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H22266
Availability details for
Quantity
  • Availability
  • Add to Cart