MAN326367504 Manchester Organics

(S)-1-Boc-3-isobutyl-piperazine

  • MDL number MFCD07772108
  • Cas Number : 928025-62-1
  • Molecular Weight : 242.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/M22270
Availability details for
Quantity
  • Availability
  • Add to Cart