MAN326367508 Manchester Organics

(S)-1-Boc-3-hydroxymethyl-piperazine

  • MDL number MFCD05863889
  • Molecular Weight : 216.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S22464

Safety Information

MSDS https://www.manchesterorganics.com/S22464
Availability details for
Quantity
  • Availability
  • Add to Cart